Learn More
NOD4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA566236
Additional Details : Weight : 0.01000kg
Description
Immunogen sequence: GLVRCFSTLQW LFRLDISFES QHILLRGDKT SRDMWATGSL PDFPAAAKFL GFRQRCIPRS LCLSECPLEP PSLTRLCATL KDCPGPLELQ LSCEFLSDQ Highest antigen sequence identity to the following orthologs - mouse 55%, rat 50%.
NOD4 is a member of the NOD (nucleotide-binding oligomerization domain) family, a group of proteins that are involved in innate immune defense. NOD4 contains a CARD-like domain, a central NOD domain and a large LRR region. NOD4, an IFN-gamma-inducible nuclear protein, plays a role in homeostatic control of innate immunity and in antiviral defense mechanisms. As a key negative regulator of NF-kappa-B and type I interferon signaling, NOD4 may be a useful target for manipulating immune responses against infectious or inflammation-associated diseases, including cancer.Specifications
NOD4 | |
Polyclonal | |
Unconjugated | |
NLRC5 | |
AI451557; AK220210; Caterpiller protein 16.1; CLR16.1; copine II; Cpne2; NLR family CARD domain containing 5; NLR family, CARD domain containing 5; Nlrc5; NOD27; NOD4; NOD-like receptor C5; Nucleotide-binding oligomerization domain protein 27; Nucleotide-binding oligomerization domain protein 4; nucleotide-binding oligomerization domain, leucine rich repeat and CARD domain containing 5; nucleotide-binding oligomerization domains 27; protein NLRC5 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
84166 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q86WI3 | |
NLRC5 | |
Recombinant Human NOD4 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
For Research Use Only