missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nodal Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA567917
Additional Details : Weight : 0.01000kg
Description
Predicted to react with Mouse and Rat samples.
NODAL is an important TGF-beta family protein in cellular differentiation. While constitutively expressed in many developing tissues, NODAL is problematically associated with the differentiation and development of many cancers. The expression of NODAL is regulated by a Notch signaling pathway, which when disrupted contributes to the aggressive behavior of metastatic cancer cells, particularly in melanoma. The abundance of NODAL expression in melanoma can help determine the stage of cancer and metastatic type/progression when tested in IHC. NODAL interactions with Cripto also predispose toward tumor progression. Overexpression of NODAL is also associated with preeclamptic placenta.Specifications
Nodal | |
Polyclonal | |
Unconjugated | |
NODAL | |
early embryo mesoderm formation; HTX5; MGC138230; Nodal; nodal growth differentiation factor; nodal homolog; Tg.413d; transgenic lethal 413.d | |
Rabbit | |
affinity chromatography | |
RUO | |
18119, 294503, 4838 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P43021, Q96S42 | |
NODAL | |
Synthesized peptide derived from human Nodal. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |