Learn More
Non Ratio Turbidity Standard, Reagecon™

Use turbidity standards for precise measurements with nephelometeric instruments. Non Ratio Turbidity Standards, Reagecon™ are composed of suspended polymer microspheres to eliminate the problems with Formazin standards. They are NIST traceable, US EPA approved and certified accurate to ±1%.
Brand: Reagecon CRS-100-100
Description
These turbidity standards remove the handling, stability and accuracy problems associated with traditional Formazin turbidity standards. They are certified traceable to NIST Reference Material 1690, and are available in a range of concentrations from 0–4000 NTU.
- Non-toxic and non-carcinogenic, with no special handling or disposal requirements
- Well defined spherical shape of particles, 0.01–0.2μm in diameter
- Particles stay in suspension—they do not deteriorate or settle out
- A long stable shelf life of 2 years at all concentrations
- Convenient, ready-to-use solution—mixing is discouraged as it entrains air
Specifications
Non Ratio Turbidity Standard | |
Nephelometric turbidity measurement | |
100 NTU | |
100 mL |
Non Ratio Instruments | |
±1% | |
NIST, US EPA Approved |
Product Suggestions
Customers who viewed this item also viewed
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
CRS10024B1 | Certificate of Analysis | 01/02/2025 | CRS-100-100 |
CRS10024G1 | Certificate of Analysis | 14/01/2025 | CRS-100-100 |
CRS10023C1 | Certificate of Analysis | 16/01/2024 | CRS-100-100 |
CRS10022D1 | Certificate of Analysis | 04/10/2022 | CRS-100-100 |
CRS10021B1 | Certificate of Analysis | 09/09/2021 | CRS-100-100 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.