missing translation for 'onlineSavingsMsg'
Learn More

Non Ratio Turbidity Standard, Reagecon™
BIOPHARMA

Use turbidity standards for precise measurements with nephelometeric instruments. Non Ratio Turbidity Standards, Reagecon™ are composed of suspended polymer microspheres to eliminate the problems with Formazin standards. They are NIST traceable, US EPA approved and certified accurate to ±1%.

Brand:  Reagecon CRS-1-100

 View more versions of this product

Product Code. 11798495

  • £75.65 / 100mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

These turbidity standards remove the handling, stability and accuracy problems associated with traditional Formazin turbidity standards. They are certified traceable to NIST Reference Material 1690, and are available in a range of concentrations from 0–4000 NTU.

  • Non-toxic and non-carcinogenic, with no special handling or disposal requirements
  • Well defined spherical shape of particles, 0.01–0.2μm in diameter
  • Particles stay in suspension—they do not deteriorate or settle out
  • A long stable shelf life of 2 years at all concentrations
  • Convenient, ready-to-use solution—mixing is discouraged as it entrains air
Specifications

Specifications

Non Ratio Turbidity Standard
Nephelometric turbidity measurement
1 NTU
100 mL
Non Ratio Instruments
±1%
NIST, US EPA Approved
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Non Ratio Turbidity Standard, Reagecon™ > Turbidity: 1 NTU; Volume: 100mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.