missing translation for 'onlineSavingsMsg'
Learn More

Micronova™ Non-Sterile BucketLiner
BIOPHARMA

Micronova™ Non-Sterile BucketLiner creates an impermeable barrier between strong disinfectants and buckets or between product and container.

Brand:  Micronova™ BKL7-3119

 View more versions of this product

Product Code. 13182370

This item is not returnable. View return policy

Description

Description

  • Laminate is 75% LDPE, 25% Nylon
  • Excellent for disinfectants and cleaning solutions
  • Ideal for product parts or raw material transfer
  • 3 mil thickness
  • Flat packed, individually double-bagged or bulk packed
  • Holds up to autoclave with liquid contents
  • Non-Sterile liners are bulk packed 50 per bag, 200 Liners per case
Specifications

Specifications

Bucket Liner
Not Autoclavable
31 x 19 in.
B-7 series Bucket or Similar Sized tote
75 cm
Non-sterile
LDPE, Nylon
Clear
Product Transport or Liquid Containment
48 cm
24/Pk.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers