missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Greiner Bio-One Non-treated 96-well Microplates
Manufactured without surface treatment to create low biomolecular binding for sensitive biochemical screening applications. Greiner Bio One™ Non-treated 96-Well Assay Microplates are available in polystyrene or polypropylene with a range of well designs.
Brand: Greiner Bio-One 650201
This item is not returnable.
View return policy
Description
- Footprint compatible with automated systems
- Can be traced to production through defined LOT number system
- Free of detectable endotoxins (0.003 EU/mL) and regularly tested using an FDA-approved kinetic turbidimetric LAL-test (Limulus Amoebocyte Assay)
- Manufactured from raw materials tested for leachables
- Free of detectable DNase, RNase and human DNA
- Non-pyrogenic
- Manufactured under DIN ISO 9001 guidelines
- Models with chimney design allow each well to stand on its owndiminishing the risk of sample carryover and cross contamination
Specifications
Alphanumeric | |
Standard | |
8 x 12 | |
Polypropylene | |
Non-sterile | |
355 μL | |
Yes | |
-196 to +121°C |
DIN ISO 9001 | |
Without Lid | |
Clear | |
96 | |
Non-treated | |
Round Bottom (U-well) | |
100/Cs. |