missing translation for 'onlineSavingsMsg'
Learn More

Greiner Bio-One Non-treated 96-well Microplates

Manufactured without surface treatment to create low biomolecular binding for sensitive biochemical screening applications. Greiner Bio One™ Non-treated 96-Well Assay Microplates are available in polystyrene or polypropylene with a range of well designs.

Brand:  Greiner Bio-One 651101

 View more versions of this product

Product Code. 10242711

  • £103.00 / Pack of 100

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Footprint compatible with automated systems
  • Can be traced to production through defined LOT number system
  • Free of detectable endotoxins (0.003 EU/mL) and regularly tested using an FDA-approved kinetic turbidimetric LAL-test (Limulus Amoebocyte Assay)
  • Manufactured from raw materials tested for leachables
  • Free of detectable DNase, RNase and human DNA
  • Non-pyrogenic
  • Manufactured under DIN ISO 9001 guidelines
  • Models with chimney design allow each well to stand on its owndiminishing the risk of sample carryover and cross contamination
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Alphanumeric
Standard
8 x 12
Polystyrene
Non-sterile
234 μL
Yes
-20°C to +60°C
DIN ISO 9001
Without Lid
Clear
96
Non-treated
Conical Bottom (V-well)
100/Cs.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers