missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Greiner Bio-One Non-treated 96-well Microplates

Click to view available options
Total Volume (Metric) Well:
234 μL
323 μL
340 μL
355 μL
382 μL
Material:
Polypropylene
Polystyrene
Well Shape:
Conical Bottom (V-well)
Flat Bottom (F-well)
Round Bottom (U-well)
Unit Size:
Pack of 100
Description
- Footprint compatible with automated systems
- Can be traced to production through defined LOT number system
- Free of detectable endotoxins (0.003 EU/mL) and regularly tested using an FDA-approved kinetic turbidimetric LAL-test (Limulus Amoebocyte Assay)
- Manufactured from raw materials tested for leachables
- Free of detectable DNase, RNase and human DNA
- Non-pyrogenic
- Manufactured under DIN ISO 9001 guidelines
- Models with chimney design allow each well to stand on its owndiminishing the risk of sample carryover and cross contamination
Specifications
Specifications
| Barcode | Alphanumeric |
| Certifications/Compliance | DIN ISO 9001 |
| Format | Standard |
| Lid | Without Lid |
| Array | 8 x 12 |
| Color | Clear |
| Material | Polypropylene |
| No. of Wells | 96 |
| Sterility | Non-sterile |
| Surface Treatment | Non-treated |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction