missing translation for 'onlineSavingsMsg'
Learn More

Nonacarbonyldiiron, 99%, Thermo Scientific Chemicals

Product Code. 11312158
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy
This item is not returnable. View return policy

It is an important reagent in organometallic chemistry and of occasional use in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15321-51-4
Molecular Formula C9H3Fe2O9
Molecular Weight (g/mol) 366.804
MDL Number MFCD00151465
InChI Key CGJSNKHFIBINMR-UHFFFAOYSA-N
Synonym iron nonacarbonyl, enneacarbonyldiiron, nonacarbonyldiiron, 1,3-diferrabicyclo 1.1.1 pentane-2,4,5-trione; hexakis carbon monoxide, diironnonacarbonyl, diironnonacarbonyl, purum fe
PubChem CID 71311424
IUPAC Name carbon monoxide;iron;iron(3+);methanone
SMILES [CH-]=O.[CH-]=O.[CH-]=O.[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[Fe].[Fe+3]

Specifications

Melting Point ∼100°C (decomposition)
Color Orange
Density 2.85 g/mL
Linear Formula Fe2(CO)9
Quantity 10 g
UN Number UN3179
Sensitivity Air Sensitive
Solubility Information Insoluble in water.
Formula Weight 363.79
Percent Purity 99%
Physical Form Crystalline
Chemical Name or Material Nonacarbonyldiiron
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Nonacarbonyldiiron
Signal Word
  • Danger
Hazard Category
  • Flammable solid Category 1
  • Acute toxicity Category 3
  • Acute toxicity Category 2
Hazard Statement
  • H228-Flammable solid.
  • H301-Toxic if swallowed.
  • H330-Fatal if inhaled.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'