missing translation for 'onlineSavingsMsg'
Learn More

Nonidet™ P40 Substitute, Ultrapure, Thermo Scientific Chemicals

£72.60 - £99.40

Chemical Identifiers

CAS 9002-93-1
Molecular Formula C16H26O2
Molecular Weight (g/mol) 250.38
MDL Number MFCD00132505
InChI Key JYCQQPHGFMYQCF-UHFFFAOYSA-N
Synonym Igepal CA to 630,Polyethylene glycol tert to octylphenyl ether
PubChem CID 5590
IUPAC Name 2-[4-(2,4,4-trimethylpentan-2-yl)phenoxy]ethanol
SMILES CC(C)(C)CC(C)(C)C1=CC=C(OCCO)C=C1
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
15875388
View Documents
Thermo Scientific
J19628.AP
500 mL Plastic bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15885388
View Documents
Thermo Scientific
J19628.K2
1 L Plastic bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Specifications

Chemical Identifiers

9002-93-1
250.38
JYCQQPHGFMYQCF-UHFFFAOYSA-N
5590
CC(C)(C)CC(C)(C)C1=CC=C(OCCO)C=C1
C16H26O2
MFCD00132505
Igepal CA to 630,Polyethylene glycol tert to octylphenyl ether
2-[4-(2,4,4-trimethylpentan-2-yl)phenoxy]ethanol

Specifications

9002-93-1
>250°C
1.061
1.061
MFCD00132505
JYCQQPHGFMYQCF-UHFFFAOYSA-N
2-[4-(2,4,4-trimethylpentan-2-yl)phenoxy]ethanol
5590
Nonidet™ P40 Substitute
Viscous Liquid
∼5°C
>110°C
C16H26O2
Igepal CA to 630,Polyethylene glycol tert to octylphenyl ether
CC(C)(C)CC(C)(C)C1=CC=C(OCCO)C=C1
250.38
ca 625
Videos
Safety and Handling

Safety and Handling

P264b-P270-P280-P301+P312-P305+P351+P338-P310-P330-P501c

H302-H318

missing translation for 'rtecsNumber' : MD0907600

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.