Learn More
Thermo Scientific™ Nonsterile Pipette Tips for Beckman™ Liquid Handling Systems
Increase pipetting performance through the superior tip straightness and low %CV of these pipette tips for automated liquid handling systems.
Brand: Thermo Scientific™ 919-252
544.71 GBP valid until 2024-12-31
Use promo code "21898" to get your promotional price.
This item is not returnable.
View return policy
Description
Increase pipetting performance with Thermo ScientificTM Nonsterile Pipette Tips for BeckmanTM Liquid Handling Systems, which undergo a rigorous 15-point quality control program to ensure consistency. They provide accurate and precise performance because of superior tip straightness and a low coefficient of variation. The pipette tips are validated on the actual workstations for which they are created.
Features:
- Available in low retention and extended length
- Lot-tested during production on actual workstation for which they are created
- Extended length tips compatible with 96-, 384- and 1536- DeepWell™ plates
- Low percentage coefficient of variation (%CV)
Specifications
250 μL | |
Non-sterile | |
Conductive | |
Beckman |
Non-filtered | |
Rack | |
Beckman Coulter Biomek FX, Beckman Coulter Biomek NX SPAN-8 | |
Certified Quality |