missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RNase-free Microfuge Tubes

Product Code. 10676825
Click to view available options
Capacity (Metric):
0.5 mL
1.5 mL
2 mL
Surface Type:
Graduated
Graduated, low-binding
Quantity:
1000 Tubes/Pk.
250 Tubes/Pk.
500 Tubes/Pk.
Unit Size:
Pack of 1000
Pack of 250
Pack of 500
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10676825

Brand: Invitrogen™ AM12450

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

These microcentrifuge tubes are guaranteed RNase- and DNase-free.

Invitrogen microcentrifuge tubes are guaranteed RNase- and DNase-free.

Features include:

  • Marked with graduations for convenience
  • Flat lids for easy labeling
  • Positive seal design that prevents caps from opening in boiling water baths
  • Autoclave-safe
TRUSTED_SUSTAINABILITY

Specifications

Color Clear
Capacity (Metric) 1.5 mL
Material Polypropylene
Quantity 250 Tubes/Pk.
Content And Storage Store at room temperature.
Product Line Invitrogen
Shipping Condition Room Temperature
Dnase Free RNase-Free, DNase-Free
Autoclavable Autoclavable
Closure Type Snap Cap
Graduated Yes
Product Type Microfuge Tube (RNase-free)
Surface Type Graduated, low-binding
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.