Learn More
Nop25 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marque: Thermo Scientific PA551958
Informations supplémentaires : Weight : 0.02000kg
Description
Immunogen sequence: HQEYLKMLAE REEALEEADE LDRLVTAKTE SVQYDHPNHT VTVTTISDLD LSGARLLGLT PPEGGAGDRS EEEASSTEKP TKALPRKSRD PLLSQRISSL TASLHAHSRK KVKRKHPRRA QDSKKPPRAP RTSKAQRRRL TGKAR Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 86%.
May bind to 28S rRNA.Spécification
Nop25 | |
Polyclonal | |
Unconjugated | |
NOL12 | |
25 kDa nucleolar protein; C78541; dJ37E16.7; Nol12; Nop25; Nucleolar protein 12; nucleolar protein of 25 kDa; nucleolar protein, 25 kDa; RGD1305327 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
79159 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9UGY1 | |
NOL12 | |
Recombinant protein corresponding to Human Nop25 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only