Learn More
uvex NORA S5 SRC Chemical-Resistant Safety Boots
NORA safety boots are characterized by very high resistance to oils, grease, fuels, alkalis, acids and disinfectants making it the ideal boot choice when working with chemicals. Boots feature a robust protective steel toe cap, with a penetration-resistant steel midsole.
Brand: uvex 9475643
26.27 GBP valid until 2024-12-31
Use promo code "21676" to get your promotional price.
This item is not returnable.
View return policy
Description
- Boots are resistant to oils, grease, fuels, alkalis, acids, and disinfectants
- Protection class S5 SRC in accordance with EN ISO 20345:2011
- A robust, protective steel to cap with a penetration-resistant steel midsole
- Free of plasticizers contained in the SVHC Candidate List of the REACH Regulation
- Toe and heel reinforcement, as well as ankle protection
- Anatomically shaped sole helps to prevent foot fatigue
- Anti-static
Specifications
Yellow | |
43 | |
Unisex | |
No | |
Safety Shoe |
PVC | |
Steel | |
Oils, grease, fuels, alkalis, acids, and disinfectants | |
S5 |