Learn More
GPE Scientific™ Norell™ Thin-Wall Transparent Fluoropolymer FEP Tubing
Manufactured from special virgin grade thermoplastic FEP fluoropolymer
Brand:  GPE Scientific™ TWT-12
 View more versions of this product
Product Code. 13190833
  • £185.00 / Pack of 4
Estimated Shipment: 27-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Unlike PTFE tubing, this material can be heat sealed
  • Heat sealing can offer extremely large possibilities for packaging and/or storage of samples (corrosives, liquids, solids, etc.)
  • Fluoropolymer tubing can also be used where snug and tight fit over glass or metal tubing is required
Specifications
Specifications
12.8 mm