Learn More
GPE Scientific™ NorLoc™ NMR Tube Caps
GPE Scientific™ NorLoc™ NMR Tube Caps can be applied easily and quickly, increasing personal safety and saving valuable time.
Brand:  GPE Scientific™ NORLOC2-5-LY-100PK
 View more versions of this product
Product Code. 15658995
  • £84.95 / Pack of 100
Estimated Shipment: 03-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Features a patented design that addresses many of the flaws in the traditional NMR tube cap.
  • Can be applied much more easily and quickly, thereby increasing personal safety and saving valuable time.
  • Incorporates an advanced seal design, conferring superior holding and sealing capabilities.
  • Superior retention of the cap to the tube forms a vaulted seal that safeguards precious or critical samples, even during refrigerated cold storage and variable temperature cycling and withstands repeated removals and reapplications
Specifications
Specifications
NMR Tube Cap
Lemon Yellow
100/Pk.
5 mm