missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Normal Goat IgG, Negative Control, R&D Systems™
Goat Polyclonal Antibody has been used in 282 publications
Brand: R&D Systems AB-108-C
115.20 GBP valid until 2025-03-28
Use promo code "25306" to get your promotional price.
This item is not returnable.
View return policy
Description
IgG Polyclonal specifically detects IgG in Goat samples. It is validated for Control.
Specifications
IgG | |
LYOPH | |
Control | |
IGHG1 | |
NA | |
1 mg | |
Serum was obtained from naive (non-immunized) goats and purified for use as normal goat IgG. | |
Goat | |
IgG |
Polyclonal | |
Unconjugated | |
Immunoglobulin G, ImmunoglobulinG | |
Goat | |
Protein A or G purified | |
3500 | |
Dissolve the lyophilized goat IgG in sterile PBS, pH 7.4. If 1 mL of PBS is used, the antibody concentration will be 1 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Normal Goat IgG, Negative Control, R&D Systems™ > 1mg, Unlabeled
Spot an opportunity for improvement?Share a Content Correction