missing translation for 'onlineSavingsMsg'
Learn More

Normal Mouse Serum, Invitrogen™

Mouse Control

Brand:  Invitrogen 016501

Product Code. 15953104

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

This serum is collected from NIH Swiss mouse.

Normal Sera are non-immune serum samples from goat, human, mouse, rabbit and other species that contain the usual complement of serum proteins, including the various classes of immunoglobulins that exist in healthy animals of the particular species. Normal serum provides a sufficient quantity of endogenous proteins to saturate and block nonspecific binding sites on fixed samples or assay substrates. When used as an antibody diluent, non-immune serum also supplies a native chemical environment for antibodies to function properly without adding foreign protein components. Normal serum is frequently used for blocking or saturating generalized binding interactions for immunodetection methods, especially those involving tissue samples such as immunohistochemistry (IHC). Normal sera are also useful as controls for testing general and specific antibody purification methods.

Order Info

Shipping condition: Wet ice

TRUSTED_SUSTAINABILITY
Specifications

Specifications

4°, C
Normal Mouse Serum
Conc. Not Determined
Control
RUO
Lyophilized
5 mL
whole serum with 0.1% sodium azide
Ig
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.