missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Normal Rabbit Serum

Normal rabbit serum.

Brand:  Invitrogen™ PLN5001

43.83 GBP valid until 2024-12-14
Use promo code "21632" to get your promotional price.


Product Code. 15574404

  • £65.75 / 5mL
Estimated Shipment: 17-12-2024
to see stock.
Explore more special offers
Add to basket

Alert:

To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21632” to get your promotional price

This item is not returnable. View return policy

Description

Description

Normal rabbit serum.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Store in freezer (-5°C to -30°C).
Liquid
5 mL
Rabbit
Rabbit Serum
Dry Ice
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers