missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eppendorf™ Nose Cone Filter
Use this tip cone filter with Eppendorf pipettes.
Brand: Eppendorf™ 3111606004
30.64 GBP valid until 2024-05-31
Use promo code "21834" to get your promotional price.
Additional Details : Weight : 0.25000kg
Description
Use this tip cone filter with Eppendorf pipettes.
- Color code: turquoise
- 10mL
- Available in packs of 50
Specifications
50Pack | |
Nose Cone Filter | |
10 mL |