missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NOTCH2 Monoclonal Antibody (605724), Invitrogen™
Rat Monoclonal Antibody
Brand: Invitrogen MA524274
Code : NEW
Additional Details : Weight : 0.00010kg
Description
Reconstitute in sterile PBS to a final concentration of 0.5 mg/mL.
NOTCH2 Monoclonal antibody specifically detects NOTCH2 in Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry.Specifications
NOTCH2 | |
Monoclonal | |
0.5 mg/mL | |
PBS with 5% trehalose and No Preservative | |
O35516 | |
NOTCH2 | |
CHO-derived recombinant mouse Notch-2 Leu26-Val528 (predicted) | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG1 |
Flow Cytometry, Immunocytochemistry | |
605724 | |
Unconjugated | |
NOTCH2 | |
AGS2; AI853703; HJCYS; hN2; Motch B; N2; N2ECD; N2ICD; Neurogenic locus notch homolog protein 2; notch 2; Notch 2 extracellular truncation; Notch 2 intracellular domain; notch gene homolog 2; Notch homolog 2; notch receptor 2; Notch2 | |
Rat | |
Protein A/G | |
RUO | |
18129 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |