missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ NotI (10 U/μL)

The NotI restriction enzyme recognizes GC^GGCCGC sites and cuts best at 37°C in O buffer (Isoschizomers: CciNI).

£43.21 - £175.00

Specifications

Concentration 10 U/μL
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10640081
View Documents Promotion Details
Thermo Scientific™
ER0592
1500 U
£175.00
1500 units
Please to purchase this item. Need a web account? Register with us today!
10440240
View Documents Promotion Details
Thermo Scientific™
ER0595
500 U
£73.50
500 units
Please to purchase this item. Need a web account? Register with us today!
10793911
View Documents Promotion Details
Thermo Scientific™
ER0591
300 U
£43.21
300 units
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Ad2 DNA digested with NotI, 0.7% agarose, 7 cleavage sites

conventional restriction endonucleases are a large collection of high quality restriction enzymes, optimized to work in one of the buffers of the Five Buffer System. In addition, the universal Tango buffer is provided for convenience in double digestions. All of the enzymes exhibit 100% activity in the recommended buffer and reaction conditions. To ensure consistent performance, Thermo Scientific restriction enzyme reaction buffers contain premixed BSA, which enhances the stability of many enzymes and binds contaminants that may be present in DNA preparations.

Highlights

  • Superior quality -stringent quality control and industry leading manufacturing process
  • Convenient color-coded Five Buffer System
  • Includes universal Tango buffer for double-digestions
  • BSA premixed in reaction buffers
  • Wide selection of restriction endonuclease specificities
Specifications

Specifications

10 U/μL
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only. Not for use in diagnostic procedures.