Learn More
Invitrogen™ NovaFluor™ Antibody Conjugation Kits
Get the most out of your panel design by conjugating antibodies to NovaFluors: dyes specifically designed for spectral hygiene to have narrow emission spectra and minimal cross-laser excitation.
£367.00 - £431.00
Product Code | Brand | Label or Dye | Product Type | Emission | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Label or Dye | Product Type | Emission | Price | Quantity & Availability | |||
17728303
|
Invitrogen™
K06T04L008 |
NovaFluor™ Blue 510 | Conjugation Kit | 510 nm |
£402.00
100µg |
Estimated Shipment: 06-02-2025
Log in to see stock. |
|||
17768293
|
Invitrogen™
K06T04L002 |
NovaFluor™ Blue 530 | Conjugation Kit | 530 nm |
£402.00
100µg |
Estimated Shipment: 06-02-2025
Log in to see stock. |
|||
17585325
|
Invitrogen™
K06T04L009 |
NovaFluor™ Blue 555 | Conjugation Kit | 555 nm |
£402.00
100µg |
Estimated Shipment: 24-01-2025
Log in to see stock. |
|||
17738303
|
Invitrogen™
K06T04L010 |
NovaFluor™ Blue 585 | Conjugation Kit | 585 nm |
£402.00
100µg |
Estimated Shipment: 06-02-2025
Log in to see stock. |
|||
17778293
|
Invitrogen™
K06T04L003 |
NovaFluor™ Blue 610/30S | Conjugation Kit | 610 nm |
£403.00
100µg |
Estimated Shipment: 06-02-2025
Log in to see stock. |
|||
17748303
|
Invitrogen™
K06T04L011 |
NovaFluor™ Blue 610/70S | Conjugation Kit | 610 nm |
£402.00
100µg |
Estimated Shipment: 06-02-2025
Log in to see stock. |
|||
17788293
|
Invitrogen™
K06T04L004 |
NovaFluor™ Blue 660/40S | Conjugation Kit | 660 nm |
£403.00
100µg |
Estimated Shipment: 06-02-2025
Log in to see stock. |
|||
17826781
|
Invitrogen™
K06T04L021 |
NovaFluor™ Blue 690 | Conjugation Kit | 690 nm |
£374.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17758303
|
Invitrogen™
K06T04L012 |
NovaFluor™ Blue 660/120S | Conjugation Kit | 660 nm |
£402.00
100µg |
Estimated Shipment: 06-02-2025
Log in to see stock. |
|||
18065331
|
Invitrogen™
K06T04L025 |
NovaFluor™ Blue 725 | Conjugation Kit | 725 nm |
£431.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
18025341
|
Invitrogen™
K06T04L026 |
NovaFluor™ Blue 760 | Conjugation Kit | 760 nm |
£430.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17708313
|
Invitrogen™
K06T04L017 |
NovaFluor™ Red 660 | Conjugation Kit | 660 nm |
£402.00
100µg |
Estimated Shipment: 06-02-2025
Log in to see stock. |
|||
17718313
|
Invitrogen™
K06T04L018 |
NovaFluor™ Red 685 | Conjugation Kit | 685 nm |
£402.00
100µg |
Estimated Shipment: 06-02-2025
Log in to see stock. |
|||
17728313
|
Invitrogen™
K06T04L019 |
NovaFluor™ Red 700 | Conjugation Kit | 700 nm |
£402.00
100µg |
Estimated Shipment: 06-02-2025
Log in to see stock. |
|||
17738313
|
Invitrogen™
K06T04L020 |
NovaFluro™ Red 710 | Conjugation Kit | 710 nm |
£402.00
100µg |
Estimated Shipment: 06-02-2025
Log in to see stock. |
|||
17846781
|
Invitrogen™
K06T04L023 |
NovaFluor™ Red 725 | Conjugation Kit | 725 nm |
£367.00
100µg |
Estimated Shipment: 06-02-2025
Log in to see stock. |
|||
17856781
|
Invitrogen™
K06T04L024 |
NovaFluor™ Red 755 | Conjugation Kit | 755 nm |
£374.00
100µg |
Estimated Shipment: 06-02-2025
Log in to see stock. |
|||
17798293
|
Invitrogen™
K06T04L005 |
NovaFluor™ Yellow 570 | Conjugation Kit | 570 nm |
£395.00
100µg |
Estimated Shipment: 06-02-2025
Log in to see stock. |
|||
17768303
|
Invitrogen™
K06T04L013 |
NovaFluor™ Yellow 590 | Conjugation Kit | 590 nm |
£402.00
100µg |
Estimated Shipment: 06-02-2025
Log in to see stock. |
|||
17708303
|
Invitrogen™
K06T04L006 |
NovaFluor™ Yellow 610 | Conjugation Kit | 610 nm |
£403.00
100µg |
Estimated Shipment: 06-02-2025
Log in to see stock. |
|||
17718303
|
Invitrogen™
K06T04L007 |
NovaFluor™ Yellow 660 | Conjugation Kit | 660 nm |
£402.00
100µg |
Estimated Shipment: 06-02-2025
Log in to see stock. |
|||
17778303
|
Invitrogen™
K06T04L014 |
NovaFluor™ Yellow 690 | Conjugation Kit | 690 nm |
£402.00
100µg |
Estimated Shipment: 06-02-2025
Log in to see stock. |
|||
17788303
|
Invitrogen™
K06T04L015 |
NovaFluor™ Yellow 700 | Conjugation Kit | 700 nm |
£402.00
100µg |
Estimated Shipment: 06-02-2025
Log in to see stock. |
|||
17798303
|
Invitrogen™
K06T04L016 |
NovaFluor™ Yellow 730 | Conjugation Kit | 730 nm |
£402.00
100µg |
Estimated Shipment: 06-02-2025
Log in to see stock. |
|||
17836781
|
Invitrogen™
K06T04L022 |
NovaFluor™ Yellow 755 | Conjugation Kit | 755 nm |
£374.00
100µg |
Estimated Shipment: 06-02-2025
Log in to see stock. |
|||
30088884
|
Invitrogen™
K06T04L027 |
NovaFluor™ UV 765 | Conjugation Kit | 800 nm |
£406.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
30089849
|
Invitrogen™
K06T04L028 |
NovaFluor™ Violet 800 | Conjugation Kit | 810 nm |
£406.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
30089504
|
Invitrogen™
K06T04L029 |
NovaFluor™ Violet 745 | Conjugation Kit | 800 nm |
£406.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
30088591
|
Invitrogen™
K06T04L030 |
NovaFluor™ Yellow 810 | Conjugation Kit | 690 nm |
£406.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
30089843
|
Invitrogen™
K06T04L031 |
NovaFluor™ Blue 725 | Conjugation Kit | 745 nm |
£406.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
30088885
|
Invitrogen™
K06T04L032 |
NovaFluor™ Violet 690 | Conjugation Kit | 765 nm |
£406.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
30089827
|
Invitrogen™
K06T04L033 |
NovaFluor™ Violet 690 | Conjugation Kit | 725 nm |
£406.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
NovaFluor Conjugation Kits provide a simple workflow to conjugate a blue, yellow-green or red excitable fluorophore to your antibody of choice in just 3 easy steps. The fluorescence-by-design Phiton platform enables the addition of parameters to your current panels in discovery research and resolves stained populations with near-zero cross excitation, with a lack of emission in the violet channels that plague other fluorophores.
NovaFluor Conjugation Kit provides features including:
- A clean and bright antibody label to increase the number of parameters on conventional and spectral flow cytometers
- Resolve populations with limited/near-zero cross excitation and the lowest spread performance of any blue, yellow-green or red excited fluorescent labels
- Simple 3-step protocol, with 60 sec. of hands-on time for conjugating 10–100 μg of antibody per label
- Up to 70% less excitation when compared to PE tandems, resulting in decreased spillover and higher resolution
3-step conjugation method
- Attach NovaFluor Linker to antibody of choice
- Remove excess NovaFluor Linker from DNA-conjugate antibody
- Anneal NovaFluor to oligo-conjugated antibody
Specifications
NovaFluor™ |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.