missing translation for 'onlineSavingsMsg'
Learn More

Biochrom Novaspec Pro Spectrophotometer

Designed for general biotech laboratory use, this spectrophotometer is lightweight and easy to use. The Biochrom Novaspec Pro Spectrophotometer features a small footprint and rugged lightweight construction, ideal for laboratories where space is at a minimum.

Brand:  Biochrom 80-2120-50

Product Code. 15591166

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Performs measurements in absorbance, % transmission and concentration
  • Includes sorted methods for Bradford, BCA, Biuret and Lowry protein quantification
  • Automatic calibration at start-up
  • No lid or moving parts, this robust instrument is easy to use, clean and store
  • Graphical display enables wavelength scans, kinetic assays and standard curves to be viewed
  • Increase measurement flexibility with the optional test tube cell holder which allows measurement of 10 to 18 mm test tubes
  • The standard cell holder accepts 10 mm glass and disposable cuvets and is easily removed for cleaning
  • Optional thermostatted cell holder is available for the added capability of measurements at 37°C
  • Visible spectrophotometer covers 325 to 1100 nm allowing you to see more of your sample
  • Ideal for basic spectrophotometric measurements, great for entry level use
  • Offers powerful onboard software that is functional in viewing and analyzing data
  • Save data on a USB and results can be analyzed later
  • PVC (Print Via Computer) software is included and allows you to transfer data directly from the instrument

Services

1 year

Specifications

Specifications

-0.3 to +2.5 A
Accepts standard 10 mm pathlength glass or disposable cuvettes. Test tube and heated cell holders are available accessories.
225 mm
Graphical, peak display with track and zoom
English, French, German, Spanish, Italian, Chinese, Japanese
90/265 V
90/265 V, 50/60 Hz, 15 VA
133 mm
Flat grating
±0.010 A or ±2.0% whichever is the greater between 0 and 1.500 A (at 546 nm)
<0.002 A (at 0 A and 500 nm)
90 with PIN number protection
±1 nm
325 mm
CE
USB PC connection with PVC software
CMOS Array
225 x 325 x 133 mm
Pulsed Tungsten Halogen
325 to 1100 nm
Single Wavelength Absorbance, Single Wavelength % Transmittance, Concentration with Factor, Concentration with Single Standard, Kinetics, Life Science Methods, Cell Density
The unit, PVC software, sample 8 Pk. of disposable cuvettes,dust cover, USB cable for PC connection, 18 V power supply with EU,UK,US and Japan power cords
<0.002 A RMS at 0 A and <0.003 A RMS at 1 A both (at 600 nm)
-0.300 to 2.500 A, 0.3 to 199%T
<1%T (at 340 nm)
±2 nm
1.6 kg
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers