missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Novex™ 10%, Tris-Glycine, Protein Gel, 20-well, w/adapters
Separation Range: 30 to 150 kd
Brand: Invitrogen™ WT0102A
Includes: One box of 10 gels, adapters
Description
- Provide reproducible separation of a wide range of proteins into well-resolved bands
- Individually packaged gels for optimal experiment design
- Compatible with most protein standards
- Usable for native and denaturing protein assays
- Do not contain SDS and can be used to run proteins in native or in denatured form
Sample Loading Volume: 25μL; 1D Well
1D Gel Electrophoresis, Protein Gel Electrophoresis, Proteins, Expression, Isolation and Analysis
Specifications
10% | |
Homogeneous Page | |
30 to 150 kDa | |
1 Box | |
13 cm |
Tris-Glycine | |
Protein | |
20-well, w/adapters | |
8 cm |
For Research Use Only. Not for use in diagnostic procedures.