missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Novex™ Value™ Tris-Glycine Mini Protein Gel, 10%, 1.0 mm, 10-well
Separation Range: 30 to 250 kd; Separation Type: Denaturing, Native; For Use With: XCell SureLock Mini
Brand: Invitrogen™ XV00100PK20
Includes: 20 precast gels
Description
Features of the Novex Value gels include:- Great balance of value and performance
- Fast run conditions - separate your proteins using constant voltage in less than 60 minutes
- Improved shelf life - store gels for at least 12 months at 4°C
- Usable for native and denaturing protein assays
Services
Up to 12 months
Specifications
Store in refrigerator (2–8°C). Shelf life up to 12 months depending on gel type. |
|
Mini Gel Tank, XCell SureLock Mini-Cell | |
Mini | |
Tris-Glycine | |
Denaturing, Native | |
Protein | |
Denaturing, Native | |
Protein Gel Electrophoresis | |
Molecular Weight | |
20/Pk. | |
Store at 4°C. Shelf life is 12 months. | |
1D Well | |
8 cm | |
8 cm |
10% Value Tris-Glycine Gels, 1.0 mm, 10-well, Box of 20 | |
10% | |
1.0 mm | |
Homogeneous Page | |
up to 25 μL | |
30 to 250 kDa | |
10-well | |
Tris-Glycine Gel | |
Novex Value™ | |
Approved for shipment at Room Temperature or on Wet Ice | |
1.0 mm | |
8 cm | |
8 cm |
For Research Use Only. Not for use in diagnostic procedures.