missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Novex™ Tricine Mini Protein Gels, 16%, 1.0 mm

Invitrogen Novex Tricine Gels provide separation of low molecular weight proteins and peptides. In this system tricine replaces glycine in the running buffer, resulting in more efficient stacking and destacking for low molecular weight proteins and higher resolution of smaller peptides.

Brand:  Invitrogen™ EC66955BOX

143.64 GBP valid until 2024-12-31
Use promo code "21651" to get your promotional price.


 View more versions of this product

Product Code. 12060116

  • £189.00 / Pack of 10
Estimated Shipment: 09-01-2025
to see stock.
Explore more special offers
Add to basket

For Research Use Only. All usage must comply with product instructions.
 

Includes: Catalog numbers for gels refer to a single box. One box contains 10 gels.
This item is not returnable. View return policy

Description

Description

Invitrogen Novex Tricine Gels provide separation of low molecular weight proteins and peptides. The Tricine system is a modification of the tris-glycine discontinuous buffer system developed by Schaegger and von Jagow (Schaegger and von Jagow, 1987) specifically for resolving peptides and low molecular weight proteins. In this system the tricine replaces the glycine in the running buffer, resulting in more efficient stacking and destacking for low molecular weight proteins and higher resolution of smaller peptides.

Features of Novex Tricine protein gels:
• Increased resolution of proteins with molecular weights as low as 2 kDa
• Improved compatibility with direct sequencing of proteins after transferring to PVDF
• Minimized protein modification due to the lower pH of the tricine buffering system

Formulation
Invitrogen Tricine gels are made with high-purity, strictly quality-controlled reagents: Tris base, HCl, acrylamide, bisacrylamide, TEMED, APS, and highly purified water. Our Tricine gels have a 4% stacking gel and do not contain SDS. The Tricine system requires SDS in sample and running buffers for best results.

Choose the right Tricine gel for your protein separation
Invitrogen Tricine gels come in three polyacrylamide concentrations of 10%, 16%, and a gradient of 10–20%. Select from our many well formats, including 10-, 12-, and 15-well. Tricine gels are formulated for denaturing gel electrophoresis applications. For optimal sample preparation, we recommend Tricine SDS Sample Buffer (LC1676) and optimal separation use Tricine SDS Running Buffer (LC1675).

For transfer of proteins to a membrane, we recommend using the Novex Tris-Glycine Transfer Buffer (LC3675) if performing a traditional wet transfer using the XCell II Blot Module (EI9051) or the Mini Blot Module (B1000). Rapid semi-dry transfer can be performed using the Invitrogen Power Blotter or rapid dry transfer using the iBlot 2 Gel Transfer Device (IB21001).

Order Info

Shipping Condition: Wet Ice

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Mini Gel Tank, XCell SureLock Mini-Cell
Mini
Tricine
Up to 15 μL
Denaturing
Molecular Weight
10 Gels/Box
Store at 2°C to 8°C. Do not freeze.
8 cm
16%
1.0 mm
Denaturing
2.5 to 200 kDa
15-well
Novex™
Wet Ice
8 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.