missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ 20X Bolt™ MOPS SDS Running Buffer

Optimized for use with Bolt™ Bis-Tris Plus gels, and are available in a variety of formats

Specifications

Chemical Name or Material Running Buffer
Concentration 20X
For Use With (Equipment) Bolt
Physical Form Liquid
Product Line Bolt
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
13226499
View Documents Promotion Details
Invitrogen™
B0001
500 mL N/A
N/A
13236499
View Documents Promotion Details
Invitrogen™
B000102
5 L N/A
N/A
Description

Description

About Bolt™ Bis-Tris Plus Gels
Bolt™ Bis-Tris Plus gels are precast polyacrylamide gels designed for optimal separation of your small- to medium-sized proteins under denaturing conditions. Similar to NuPAGE™ Bis-Tris gels, Bolt™ Bis-Tris Plus gels are designed to deliver consistent gel performance and provide a neutral pH environment that minimizes protein modifications. Bolt™ gels are ideal for western blot transfer and analysis, and any other techniques where protein integrity is crucial. Also use Bolt™ gels to obtain optimal results for your day-to-day protein separation needs. Bolt™ Bis-Tris Plus gels come in four gel types and multiple well formats. A Mini Gel Tank is required for use of all Bolt™ gels.

Bolt™ gel run times may vary depending upon the type of running buffer used.

Order Info

Shelf-life is 6 months, when stored properly.

Specifications

Specifications

Running Buffer
Bolt
Bolt
20X
Liquid
Videos
Safety and Handling

Safety and Handling

Recommended Storage : MOPS SDS Running Buffer (20X).

Store at room temperature.

Shelf-life is 6 months, when stored properly.

SDS
Documents

Documents

Product Certifications

For Research Use Only. Not for use in diagnostic procedures.