missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ 4X Bolt™ LDS Sample Buffer

Optimized for use with Bolt™ Bis-Tris Plus gels, and are available in a variety of formats

£17.83 - £149.00

Specifications

Content And Storage Sample Buffer (4X) containing lithium dodecyl sulfate (LDS) at pH 8.5 with SERVA Blue G250 and phenol red.

Store at 4°C to 25°C.

Buffer Sample Loading Buffers
Concentration 4X
For Use With (Equipment) Bolt Mini Gel Tank
Gel Type Bis-Tris Plus
View More Specs

Includes: lithium dodecyl sulfate (LDS) at pH 8.4.

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
13276499
View Documents
Invitrogen™
B0007
10 mL
£17.83
10mL
Please to purchase this item. Need a web account? Register with us today!
13286499
View Documents
Invitrogen™
B0008
250 mL
£149.00
250mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Novex™ Bolt™ buffers and sample reducing agents are optimized for use with Bolt Bis-Tris Plus gels, and are available in a variety of formats.

About Bolt™ Bis-Tris Plus Gels
Bolt™ Bis-Tris Plus gels are precast polyacrylamide gels designed for optimal separation of your small- to medium-sized proteins under denaturing conditions. Similar to NuPAGE™ Bis-Tris gels, Bolt™ Bis-Tris Plus gels are designed to deliver consistent gel performance and provide a neutral pH environment that minimizes protein modifications. Bolt™ gels are ideal for western blot transfer and analysis, and any other techniques where protein integrity is crucial. Also use Bolt™ gels to obtain optimal results for your day-to-day protein separation needs. Bolt™ Bis-Tris Plus gels come in four gel types and multiple well formats. A Mini Gel Tank is required for use of all Bolt™ gels.

Bolt™ gel run times may vary depending upon the type of running buffer used.

Specifications

Specifications

Sample Buffer (4X) containing lithium dodecyl sulfate (LDS) at pH 8.5 with SERVA Blue G250 and phenol red.

Store at 4°C to 25°C.

4X
Bis-Tris Plus
Bolt™ Bis-Tris Plus Gels
Sample Loading Buffers
Bolt Mini Gel Tank
LDS Sample Buffer
Bolt™
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ 4X Bolt™ LDS Sample Buffer

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.