missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novex™ Novex™ AP Chemiluminescent Substrate (CDP-Star™)
Novex™ AP Chemiluminescent Substrate (CDP-Star™)
Brand: Novex™ WP20002
Code : 50
Additional Details : Weight : 0.50000kg
Description
Novex™ AP Chemiluminescent Substrate is a non-radioactive ready-to-use solution of CDP-Star for chemiluminescence-based immunodetection of alkaline phosphatase (AP) on western blot or dot blot membranes. This reagent provides detection sensitivities superior to that of precipitating chromogenic substrates. Low picogram levels of detection can be achieved using either X-ray film or imaging equipment.Specifications
Single bottle labeled: Novex™ AP Chemiluminescent Substrate (CDP-Star™). Store at 4°C. |
|
Liquid | |
Novex™ | |
CDP-Star™ | |
AP (Alkaline Phosphatase) Substrate |
Chemiluminescent | |
Southern, Northern, and Western Blotting | |
1 bottle | |
Chemical Substrate | |
Approved for shipment at Room Temperature or on Wet Ice |
For Research Use Only. Not for use in diagnostic procedures.