missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Novex™ AP Mouse Chemiluminescent Detection Kit

Provides superior chemiluminescent detection of proteins transferred to either nitrocellulose or PVDF membranes and treated with a protein-specific primary antibody

Brand:  Invitrogen™ SLF1021

Product Code. 15130839

  • £157.00 / Each
Estimated Shipment: 02-01-2025
to see stock.
Explore more special offers
Add to basket

Includes: Secondary AP-conjugated anti-mouse antibody, CDP-Star™ chemiluminescent substrate, and an enhancer, sufficient for 10 mini-membranes processed using the iBind™ Western System
This item is not returnable. View return policy

Description

Description

Detection is performed by treatment of the membrane with an alkaline phosphatase (AP)-conjugated anti-mouse secondary antibody followed by a ready-to-use chemiluminescent CDP-Star™ substrate of alkaline phosphatase. Protein-specific signal is then captured by a chemiluminescent-compatible imaging system or X-ray film.

The Novex™ AP Mouse Chemiluminescent Detection Kit is compatible with most western protocols but was designed for use with the iBind™ Western System.

  • Easy-to-use protocol
  • High specificity, clean background
  • Ultra-sensitivity
  • Long-lasting signals - up to 5 days
  • Results in less 5 minutes

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Store at 2–4°C.
Kit
Novex™
CDP-Star AP substrate
iBind™
1 kit
Protein
AP (Alkaline Phosphatase) Substrate
Chemiluminescence
Western Blotting
Mouse
10 Mini Membranes
Secondary AP-conjugated anti-mouse antibody, CDP-Star™ chemiluminescent substrate, and an enhancer
Mouse
Store at 2° to 4°C.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.