Learn More
Invitrogen™ Novex™ AP Rabbit Chemiluminescent Detection Kit
Provides superior chemiluminescent detection of proteins transferred to either nitrocellulose or PVDF membranes and treated with a protein-specific primary antibody
Brand: Invitrogen™ SLF1022
Code : NEW
Additional Details : Weight : 0.02000kg
Includes: Secondary AP-conjugated anti-rabbit antibody, CDP-Star™ chemiluminescent substrate, and an enhancer, sufficient for 10 mini-membranes processed using the iBind™ Western System
Description
Detection is performed by treatment of the membrane with an alkaline phosphatase (AP)-conjugated anti-rabbit secondary antibody followed by a ready-to-use chemiluminescent CDP-Star™ substrate of alkaline phosphatase. Protein-specific signal is then captured by a chemiluminescent-compatible imaging system or X-ray film.
The Novex™ AP Rabbit Chemiluminescent Detection Kit is compatible with most western protocols but was designed for use with the iBind™ Western System.
- Easy-to-use protocol
- High specificity, clean background
- Ultra-sensitivity
- Long-lasting signals - up to 5 days
- Results in less 5 minutes
Specifications
Store at 2–4°C. | |
Kit | |
Novex™ | |
CDP-Star AP substrate | |
iBind™ | |
Rabbit | |
Store at 2° to 4°C. |
Chemiluminescent | |
Western Blotting | |
Rabbit | |
10 Mini Membranes | |
Secondary AP-conjugated anti-rabbit antibody, CDP-Star™ chemiluminescent substrate, and an enhancer | |
Protein | |
AP (Alkaline Phosphatase) Substrate |
For Research Use Only. Not for use in diagnostic procedures.