Learn More
Invitrogen™ ELISA Assay Buffer (5X)
For use with antibody pairs (ELISA)
Brand: Invitrogen™ DS98200
73.86 GBP valid until 2025-05-31
Use promo code "25977" to get your promotional price.
Description
Concentrated 5X Assay Buffer for Antibody Pairs is intended to be used as:
- Blocking reagent for ELISA plates
- Diluent for ELISA standards and samples
- Diluent for the detection antibody
- Diluent for the HRP conjugate
Build Your Own Immunoassay, Cell Analysis, Immunoassays
Order Info
Shipping Condition: Wet Ice

Specifications
ELISA | |
Buffer. Store at 2-8°C. | |
Wet Ice |
200 mL | |
Concentrated |
Safety and Handling
- ELISA Assay Buffer (5X)
Signal Word
- Warning
Hazard Category
- Skin sensitiser Category 1
Hazard Statement
- H317-May cause an allergic skin reaction.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P272-Contaminated work clothing should not be allowed out of the workplace.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P362+P364-Take off contaminated clothing and wash it before reuse.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : 5-chloro-2-methyl-3(2H)-isothiazolone, 2-methyl-3(2H)-isothiazolone
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
441945-000 | Certificate of Analysis | 03/04/2025 | |
445706-000 | Certificate of Analysis | 01/04/2025 | |
437798-000 | Certificate of Analysis | 13/03/2025 | |
437494-000 | Certificate of Analysis | 10/03/2025 | |
437494-000 | Certificate of Analysis | 25/01/2025 | DS98200 |
Special Offers
-
Multiple Manufacturers
Powering Proteomics
Advance your proteomics research with essential toolsPromo Code: 25977Expires: 2025-05-31
Days
56Hours
15Minutes
36Seconds
34New Offer Save Up to 92%
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.