missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ BenchMark™ Protein Ladder

Molecular weight standard for SDS-PAGE

Brand:  Invitrogen™ 10747012

209.02 GBP valid until 2025-06-30
Use promo code "24156" to get your promotional price.


Product Code. 11558636

  • £234.00 / Pack of 2

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers

For Research Use Only. All usage must comply with product instructions.
 

Includes: Two vials of 250μL each provided in loading buffer consisting of 50mM Tris-HCl (pH 6.8), 2mM EDTA, 10mM DTT, 2% (w/v) SDS, 10% (w/v) glycerol, 0.01% (w/v) bromophenol blue
This item is not returnable. View return policy

Description

Description

The BenchMark Protein Ladder consists of 15 recombinant proteins ranging in molecular weight from 10 to 220kDa and can be used with NuPAGE™,Tris-Glycine, or Tricine gels.

  • Extra-intense 20- and 50-kDa orientation bands to ensure proper identification of each protein
  • Affinity-purified proteins to provide sharp bands and clear background
  • A ready-to-use format for direct application to an SDS-PAGE ge
  • Detection with Coomassie™ Brilliant Blue R-250, silver, or other protein stain
  • Sufficient reagent for 100 mini gels or 50 regular gels using Coomassie Brilliant Blue R-250
  • Gel Compatibility: Bolt Bis-Tris Plus Gels, NovexTricine Gels, NovexTris-Glycine Gels, NuPAGE Bis-Tris Gels, NuPAGE Tris-Acetate Gels, SDS-PAGE Gels

1D Gel Electrophoresis, Protein Gel Electrophoresis, Protein Gel Staining and Imaging, Proteins, Expression, Isolation and Analysis

Order Info

Shipping Condition: Approved for shipment on Wet or Dry Ice

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Two vials of 250 μL each are provided in loading buffer consisting of 50 mM Tris-HCl (pH 6.8), 2 mM EDTA, 10 mM DTT, 2% (w/v) SDS, 10% (w/v) glycerol, 0.01% (w/v) bromophenol blue.

Store at -20°C. Avoid repeated freezing and thawing.
Protein Ladder
10 to 220 kDa
Bolt™ Bis-Tris Plus Gels, Novex™ Tricine Gels, Novex™ Tris-Glycine Gels, NuPAGE™ Bis-Tris Gels, NuPAGE™ Tris-Acetate Gels, SDS-PAGE Gels
BenchMark™
Approved for shipment on Wet or Dry Ice
15
Yes
Unstained
220, 160, 120, 100, 90, 80, 70, 60, 50, 40, 30, 25, 20, 15, 10 kDa
2 x 250 μL
Western Blotting, SDS-PAGE
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

  • Multiple Manufacturers
    Big discounts are available on a range of popular products including chemicals, lab essentials, equipment, consumables, life science and safety products.
    Promo Code: 24156

    Expires2025-06-30

    Hot Deal

    *Discount will apply to qualifying orders received by Fisher Scientific between 01/05/2025 and 30/06/2025. Promotion code must be presented at the time of order. Discount applies to the list price in effect at the time the order is received. Promotion cannot be combined with other discounts or promotions. Offer void where prohibited, licensed, or restricted by federal, state, provincial, or local laws or regulation or agency/institutional policy. Other restrictions may apply.

    Terms & Conditions See Full Promo Details
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ BenchMark™ Protein Ladder > 2 x 250uL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.