missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Blotting Roller

Compatible with E-PAGE™, Novex Mini Gels, and NuPAGE™ Gels

Brand:  Invitrogen™ LC2100

21.94 GBP valid until 2024-12-31
Use promo code "21745" to get your promotional price.


Product Code. 10505003

  • £24.12 / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket

For Research Use Only. All usage must comply with product instructions.
 

Includes: One blotting roller
This item is not returnable. View return policy

Description

Description

Designed for easy and efficient removal of bubbles when assembling gel/membrane sandwiches for blotting both E-PAGE™ gels and mini-gels such as NuPAGE™ and Novex™ Tris-Glycine gels. Roller is 8.6cm wide and consists of a Delrin roller attached to a stainless steel handle.

Proteins, Expression, Isolation and Analysis, Western Blotting

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY
Specifications

Specifications

One blotting roller.
E-PAGE™ Gel, Novex™ Mini Gel, NuPAGE™ Gel
Blotting Roller
Room Temperature
Western Blot
Novex
1 unit
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.