Learn More
Invitrogen™ Bolt™ Antioxidant
Description
Requires
A Bolt™ Mini Gel Tank is required for use of all Bolt gels.
Bolt Antioxidant migrates with reduced proteins to maintain reducing conditions and to prevent re-oxidation of sensitive amino acids such as methionine and tryptophan. Bolt Antioxidant must be used with samples reduced using Bolt Sample Reducing Agent or other reducing agent.
- Optimized for neutral pH of Bolt Bis-Tris Plus gels and is not compatible with other gel chemistries
- Can also be used with Bolt Transfer Buffer to enhance transfer of reduced proteins to membranes when using Mini Blot Module
1D Gel Electrophoresis, Protein Gel Electrophoresis, Proteins, Expression, Isolation and Analysis, Western Blotting
Specifications
Specifications
| Product Type | Antioxidant |
| Buffer | Antioxidant |
| Content And Storage | Store at 4°C. |
| Form | Liquid |
| Gel Compatibility | Bolt Bis-Tris Plus Gel |
| Product Line | Bolt |
| Quantity | 15 ml |
Safety and Handling
- 15ML BOLT ANTIOXIDANT
- Danger
- Serious eye damage/eye irritation Category 2
- Reproductive toxicity Category 1B
- H319-Causes serious eye irritation.
- H360-May damage fertility or the unborn child.
- P201-Obtain special instructions before use.
- P202-Do not handle until all safety precautions have been read and understood.
- P264-Wash hands thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : Dimethylformamide
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.