Learn More
Invitrogen™ Mini Blot Module
Wet transfer device used exclusively with the Bolt Minigel Tank
Brand: Invitrogen™ B1000
317.05 GBP valid until 2024-12-31
Use promo code "21658" to get your promotional price.
Includes: Bolt Mini Blot Module with Gasket; Blotting Sponge Pads (4); Blotting Tweezer; Blotting Roller
Description
The Bolt Minigel Tank accommodates one blot module per chamber, or two blot modules total with the side-by-side layout. The Bolt Mini Blot Module is compatible with Bolt Bis-Tris Plus gels and accommodates one gel per wet transfer. The universal connection and molded gasket make the blot module easy to use, while the inner core of the blot module allows for use of less methanol-based transfer buffer per wet transfer. At recommended conditions and constant voltage, proteins can be transferred to nitrocellulose or PVDF membranes typically in 30 to 60 minutes.
- Capacity: ≤ 2 Blot Modules/Minigel Tank, 1 Minigel/Blot Module
About Bolt Bis-Tris Plus Gels
- Bolt Bis-Tris Plus gels are precast polyacrylamide gels designed for optimal separation of small- to medium-sized proteins under denaturing conditions
- Similar to NuPAGE™ Bis-Tris gels, Bolt Bis-Tris Plus gels are designed to deliver consistent gel performance and provide a neutral pH environment that minimizes protein modifications
- Bolt gels are ideal for Western blot transfer and analysis, and any other techniques where protein integrity is crucial; also use Bolt gels to obtain optimal results for day-to-day protein separation needs
- Bolt Bis-Tris Plus gels come in four gel types and multiple well formats
- Each box contains 10 gels
- Bolt Mini Gel Tank is required for use of all Bolt gels
1D Gel Electrophoresis, Protein Gel Electrophoresis, Proteins, Expression, Isolation and Analysis, Western Blotting
Specifications
Mini Blot Module Blotting Tweezer Blotting Roller 4 Blotting Sponges |
|
Wet Transfer | |
Mini Precast Protein Gels | |
Nitrocellulose, PVDF | |
Blot Module | |
1 unit |
≤2 Blot Modules/Mini-Gel Tank, 1 Mini-Gel/Blot Module | |
Mini Gel Tank | |
Mini (8 x 8 cm) | |
Wet | |
Energy efficient, Less hazardous | |
Vertical |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.