Learn More
Invitrogen™ CRP Human ELISA Kit
Sandwich ELISA Kit
Brand: Invitrogen™ KHA0031
434.03 GBP valid until 2024-11-29
Use promo code "21683" to get your promotional price.
Includes: Pre-coated plate, standard, detector antibody, Streptavidin-HRP, diluents, wash buffer, chromogen, stop solution, and plate covers
Description
The Human C-Reactive Protein(Hu CRP) ELISA quantitates Hu CRP in human serum and plasma. The assay will exclusively recognize both natural and recombinant Hu CRP. Principle of the method The Human CRP solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
CRP (C-reactive protein) is a major cyclic, pentameric acute phase protein compound consisting of five identical, noncovalently bound, nonglycosylated subunits. Eeach subunit is made up of 206 amino acids, and has a molecular weight of 24 kDa. CRP is produced by the liver, and its plasma levels rise dramatically during inflammatory processes occurring in the body. CRP is an initiator of classical complement cascade, binds to several nuclear components (chromatin, histones, etc.), plays an important role in innate immunity, and mediates activities associated with pre-immune nonspecific host resistance. Specifically, there is a 100-1000 fold increase of CRP in response to injury, infection or inflammation. CRP is opsonic, an initiator of the classical complement cascade, and an activator of monocytes/macrophages. CRP also binds to several nuclear components including chromatin, histones and snRNP, suggesting that it may play a role as a scavenger during cell necrosis. CRP is involved in several host defense related functions based on its ability to recognize foreign pathogens and damaged cells of the host, and initiates pathogen elimination by interacting with humoral and cellular effector systems in the blood. Patients with elevated basal levels of CRP have been shown to be an increased risk for hypertension and cardiovascular disease.Order Info
Shipping Condition: Dry Ice
Specifications
P02741 | |
<10 pg/mL | |
ELISA Kit | |
Human | |
Colorimetric Microplate Reader | |
PTX1 | |
9.7% | |
Pre-coated 96 well plate, Standard, Standard Dilution Buffer, Biotinylated Detection Antibody, Streptavidin-HRP, HRP Diluent, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
CRP | |
RUO | |
2°C to 8°C | |
1 hr. 20 min. |
18.75-1200 pg/mL | |
Biotin | |
Plasma, Serum | |
Colorimetric Detection | |
1401 | |
4 hr. | |
7% | |
HRP | |
96 Tests | |
Plasma, 5 μL; Serum, 5 μL | |
Human | |
4 hr. |
Safety and Handling
- Human CRP ELISA Kit
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
- H302-Harmful if swallowed.
- H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P264-Wash hands thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P273-Avoid release to the environment.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P330-Rinse mouth.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : SODIUM AZIDE
For Research Use Only. Not for use in diagnostic procedures.