Learn More
Invitrogen™ Denaturing Cell Extraction Buffer
Suitable for use in ELISA, Western blotting, sample/assay prep
Brand: Invitrogen™ FNN0091
Code : 50
Additional Details : Weight : 0.50000kg
Description
Denaturing Cell Extraction Buffer can be used for Invitrogen phosphoELISAs (i.e., ERK, JNK) that require sample treatment or boiling. After lysing cells with Denaturing Cell Extraction Buffer, lysates will not require sample treatment or boiling.
Build Your Own Immunoassay, Cell Analysis, Cell Lysis, Immunoassays, Protein Sample Preparation and Protein Purification, Proteins, Expression, Isolation and Analysis
Order Info
Shipping condition: Dry ice
Specifications
Store in freezer (-5 to -30°C). | |
Cell Extraction Buffer | |
Dry Ice |
For Research Use Only. Not for use in diagnostic procedures.