Learn More
Invitrogen™ DryEase™ Mini Cellophane
Component of DryEase Mini-Gel Drying System
Brand: Invitrogen™ NC2380
Code : 50
Additional Details : Weight : 0.00454kg
Description
Mini-Gel Drying System is a simple, easy-to-use kit for drying gels evenly using passive evaporation. The kit allows gels to be assembled on a base between two open frames, so that both outer surfaces of the gels are exposed to air. Consequently, drying is even and cracking is minimized. The proprietary Gel-Dry™ Drying Solution modulates the rate of water release to ensure crack-free gels. With the DryEase™ System, gels are dried overnight with uniform tension on all points of the gel to eliminate cracking. Gels dried with the DryEase™ System are suitable for fluorography, densitometry, autoradiography, or permanent gel storage.
Gel Drying, Protein Gel Electrophoresis, Proteins, Expression, Isolation and Analysis
Order Info
Shipping condition: Room Temperature
Specifications
Store at room temperature. | |
Mini Cellophane | |
Novex™ | |
Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.