Learn More
Invitrogen™ DryEase™ Mini-Gel Drying System
For use with DryEase™ Mini-Gel Drying System
Brand: Invitrogen™ NI2387
Code : 50
Additional Details : Weight : 0.00500kg Transport : UN number : 1170 Chem class : 3 Pack group : III
Description
Gel Drying, Protein Gel Electrophoresis, Proteins, Expression, Isolation and Analysis
Specifications
The DryEase™ Mini-Gel Drying Kit contains 2 DryEase™ Mini-Gel Drying Frames with 8 Gel-Dry™ Clamps, a DryEase™ Mini-Gel Drying Base, DryEase™ Mini Cellophane (200 pre-cut sheets), and Gel-Dry™ Drying Solution (500 ml). Store the solution at room temperature. Guaranteed stable for one year when properly stored. | |
DryEase Mini-Gel Drying System | |
Mini Gel Drying System | |
DryEase™, Novex™ |
Safety and Handling
- DryEase Mini-Gel Drying System
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 3
- Specific target organ toxicity after single exposure Category 1
Hazard Statement
- H226-Flammable liquid and vapour.
- H370-Causes damage to organs.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P233-Keep container tightly closed.
- P240-Ground/bond container and receiving equipment.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P242-Use only non-sparking tools.
- P243-Take precautionary measures against static discharge.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash hands thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P370+P378-In case of fire: Use dry sand, dry chemical or alcohol-resistant foam to extinguish.
- P403+P235-Store in a well-ventilated place. Keep cool.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : methanol, ethanol, isopropanol
For Research Use Only. Not for use in diagnostic procedures.