missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DryEase™ Mini-Gel Drying Frame
For use with DryEase™ Mini-Gel Drying System
Brand: Invitrogen™ NI2380
Code : 50
Additional Details : Weight : 0.00454kg
Description
Gel Drying, Protein Gel Electrophoresis, Proteins, Expression, Isolation and Analysis
Specifications
Store at room temperature. | |
Mini Gel Drying Frame | |
Novex™ | |
Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.