Learn More
Invitrogen™ Gel-Dry™ Clamp
For use with DryEase™ Mini-Gel Drying System
Brand: Invitrogen™ NI2100
Code : 50
Additional Details : CAS Number : 53123-88-9 Weight : 0.02000kg
Description
Gel Drying, Protein Gel Electrophoresis, Proteins, Expression, Isolation and Analysis
Specifications
Store at room temperature. | |
Gel Clamp | |
Novex™ | |
Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.