Learn More
Invitrogen™ E-PAGE™ Loading Buffer 1
For use with the E-PAGE High-Throughput Protein Electrophoresis System
Brand: Invitrogen™ EPBUF01
Code : 50
Additional Details : Weight : 0.00450kg
Description
- Compatible with E-PAGE gels
- Size: 4.5mL
1D Gel Electrophoresis, Protein Gel Electrophoresis, Proteins, Expression, Isolation and Analysis
Order Info
Shipping Condition: Room Temperature
Specifications
Each E-PAGE™ 96 kit includes 8 gels, E-PAGE™ loading buffer 1, 4.5 ml loading buffer, E-PAGE™ buffer 2, 4.5 ml SDS-free loading buffer, 4.5 ml loading buffer, and a Butterfly Opener. The E-Base™ mother and daughter devices and loading buffers are available separately. Store at room temperature. Guaranteed stable for 6 months after manufacturing when stored properly. | |
E-PAGE | |
E-PAGE™ Gels | |
Room Temperature |
Sample Loading Buffers | |
Loading Buffer | |
E-PAGE™ |
For Research Use Only. Not for use in diagnostic procedures.