missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TNF alpha Human ELISA Kit

Sandwich ELISA Kit

Brand:  Invitrogen™ KAC1751

Code : 50

Additional Details : Weight : 0.50000kg

Product Code. 10583385

  • £477.00 / 96 tests
Estimated Shipment: 11-07-2024
to see stock.
Add to basket

For Research Use Only. All usage must comply with product instructions.
 

Includes: Pre-coated plate, standard, controls, antibody-HRP, diluents, wash buffer, chromogen, stop solution, and plate covers.

Description

Description

The Human Tumor Necrosis Factor alpha (Hu TNF-α) ELISA quantitates Hu TNF-α in human serum, plasma, buffered solution, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu TNF-α. Principle of the method The Human TNF-α solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

Tumor Necrosis Factor is a polypeptide cytokine produced by monocytes and macrophages. It functions as a multipotent modulator of immune response and further acts as a potent pyrogen. TNF-alpha circulates throughout the body responding to stimuli (infectious agents or tissue injury), activating neutrophils, altering the properties of vascular endothelial cells, regulating metabolic activities of other tissues, as well as exhibiting tumoricidal activity by inducing localized blood clotting. TNF-alpha also inhibits lipoprotein lipase activity resulting in cachexia, a physical wasting condition. Activation of B-cells by the Epstein Barr virus can be inhibited by TNF-alpha. Due to its varied actions throughout the immune system, TNF-alpha may play a role in the pathogenesis of many disease states. TNF-alpha production is mediated by the action of lymphokines and endotoxins on the macrophage. Purified monocytes produce TNF-alpha within four hours of stimulation by recombinant IL-2 and there is some in vitro evidence to suggest that TNF-alpha is expressed at high levels and with prolonged kinetics in T cells stimulated by both CD2 and CD28. Secretion of TNF-alpha is enhanced by gamma interferon. TNF then induces or enhances the specific production of Class I MHC antigen, GM-CSF, and IL-1.

Order Info

Shipping Condition: Wet ice

Specifications

Specifications

P01375
3 pg/mL
ELISA Kit
Human
Colorimetric Microplate Reader
DIF-alpha,TNFA,TNFSF2,TNLG1F,TNF
3%
HRP
96 Tests
Plasma, 200 μL; Serum, 200 μL; Supernatant, 200 μL
Human
4 hr. 30 min.
15-1500 pg/mL
HRP
Plasma, Serum, Supernatant
Colorimetric
7124
6%
Pre-coated 96 well plate, Standard, Sample diluent, Incubation Buffer, HRP conjugated Detection Antibody, Conjugate Diluent, Wash Buffer, Chromogen, Stop Solution, Controls
3pg/mL
RUO
2°C to 8°C
1 hr. 20 min.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.