Learn More
Invitrogen™ HiMark™ Unstained Protein Standard
Designed for analysis of high molecular weight proteins on Novex™ NuPAGE™ Tris-Acetate Gels
Brand: Invitrogen™ LC5688
354.35 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Code : 50
Additional Details : Weight : 0.02000kg
Description
- Comprehensive - Contains markers with a wide range of high molecular weights, providing nine protein bands in the range of 30-460kDa
- Convenient - A ready-to-use formulation eliminates the need to heat, reduce, or add sample buffer prior to use
- Compatible - Intended for use with NuPAGE Novex 3-8% and 7% Tris-Acetate Gels under denaturing conditions
- Can also be used with NuPAGE Novex 4-12% Bis-Tris Gels with NuPAGE MOPS Running Buffer and Novex 4% Tris-Glycine Gel, however to obtain best results with HMW proteins, always use NuPAGE Novex Tris-Acetate Gels
HiMark High Molecular Weight Protein Standard also available in a pre-stained format.
1D Gel Electrophoresis, Protein Gel Electrophoresis, Protein Gel Staining and Imaging, Proteins, Expression, Isolation and Analysis
Order Info
Shipping Condition: Dry Ice
Specifications
Contents: one vial of 250 μL each Storage buffer: 250 mM Tris-HCl (pH 8.5), 0.5 mM EDTA, 50 mM DTT, 10% glycerol, 2% LDS, 0.2 mM Coomassie G-250, and 0.175 mM Phenol Red Storage: Upon receipt store at -20°C |
|
Protein Ladder | |
Tris-acetate gels | |
Unstained | |
500, 290, 240, 160, 116, 97, 66, 55, 40 kDa | |
250 μL | |
Western Blotting, SDS-PAGE |
9 | |
Yes | |
40 to 500 kDa | |
Bolt™ Bis-Tris Plus Gels, Novex™ Tricine Gels, Novex™ Tris-Glycine Gels, NuPAGE™ Bis-Tris Gels, NuPAGE™ Tris-Acetate Gels, SDS-PAGE Gels | |
HiMark™ | |
Dry Ice |
For Research Use Only. Not for use in diagnostic procedures.