Learn More
Invitrogen™ iBlot™ DNA Transfer Stack
Quick and efficient system for transferring DNA from agarose or polyacrylamide gels in seven minutes
Brand: Invitrogen™ IB801001
Code : 50
Additional Details : Weight : 0.50000kg
Includes: Each iBlot DNA Transfer Stack contains sponges, cathode and anode electrodes and a nylon membrane, preassembled and primed with a proprietary formulation for optimum transfer of DNA from the gel to the membrane. Transfer stacks are offered in 10-pack sizes.
Description
Unlike capillary, vacuum, or electroblotting, dry blotting requires no additional buffers or liquids resulting in less result variability.
- Time savings—transfer DNA in 7 minutes instead of 16 hours
- Reproducibility—consistent blot preparation and running conditions
- Convenience—avoid messy buffers and laborious prep work
- Consists of a Gel Transfer Device and Transfer Stacks
- Transfer stacks are preassembled and intended for single use only
- Pore size: 0.2μm
- Gel size: Midi (8 x 13cm)
DNA and RNA Purification and Analysis, Nucleic Acid Gel Electrophoresis and Blotting, Southern Blotting
Order Info
Shipping Condition: Room temperature
Specifications
Each iBlot™ DNA Transfer Stack contains sponges, cathode and anode electrodes and a nylon membrane, pre-assembled and primed with a proprietary formulation for optimum transfer of DNA from the gel to the membrane. Transfer stacks are offered in 10-pak sizes. Store iBlot™ DNA Transfer Stacks at room temperature. This product is guaranteed stable for 2 months when properly stored. | |
Polyacrylamide Gel, Agarose Gel | |
Nylon | |
iBlot | |
Sustainable disposal | |
0.2 micron |
iBlot | |
Midi (8 x 13 cm) | |
Dry | |
DNA Transfer Stack | |
Nylon Membrane | |
Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.