Learn More
Invitrogen™ Novex™ IEF Anode Buffer (50X)
An optimized pre-mixed IEF anode buffer
Brand: Invitrogen™ LC5300
Code : 50
Additional Details : Weight : 0.11000kg Transport : UN number : 1760 Chem class : 8 Pack group : II
Description
Novex IEF Anode Buffer (50X) is optimized pre-mixed IEF anode buffer for using with Novex IEF gels. IEF is a sensitive technique which is affected by many factors. The optimized pre-mixed IEF buffers reduce variability and ensure consistent results.
Isoelectric Focusing, Protein Gel Electrophoresis, Proteins, Expression, Isolation and Analysis
Order Info
Shipping Condition: Room Temperature
Specifications
Store at room temperature. | |
50 X | |
IEF Anode Buffer | |
Novex™ |
Anode & Cathode Buffers | |
IEF | |
Novex™ IEF Gels | |
Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.