missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ IEF Cathode Buffer pH 3-10 (10X)

An optimized pre-mixed IEF cathod buffer

Brand:  Invitrogen™ LC5310

78.81 GBP valid until 2024-12-20
Use promo code "21651" to get your promotional price.


Product Code. 11529076

  • £111.00 / 125mL

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers

For Research Use Only. All usage must comply with product instructions.
 
This item is not returnable. View return policy

Description

Description

IEF Cathode Buffer pH 3-10 (10X) is optimized pre-mixed IEF cathode buffer for using with Novex pH 3-10 IEF gels. IEF is a sensitive technique which is affected by many factors. The optimized pre-mixed IEF buffers reduce variability and ensure consistent results.

Isoelectric Focusing, Protein Gel Electrophoresis, Proteins, Expression, Isolation and Analysis

Order Info

Shipping Condition: Wet Ice

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Store in refrigerator (2–8°C).
10X
3 to 10
Novex™ IEF Gels
Wet Ice
Anode & Cathode Buffers
IEF
IEF Cathode Buffer
125 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.