Learn More
Invitrogen™ Novex™ IEF Cathode Buffer pH 3-7 (10X)
Optimized pre-mixed IEF buffers reduce variability and ensure consistent results
Brand: Invitrogen™ LC5370
Code : 50
Additional Details : Weight : 0.00454kg
Description
Novex IEF Cathode Buffer pH 3-7 (10X) is optimized pre-mixed IEF cathod buffer for using with Novex pH 3-7 IEF gels. IEF is a sensitive technique which is affected by many factors. The optimized pre-mixed IEF buffers reduce variability and ensure consistent results.
Isoelectric Focusing, Protein Gel Electrophoresis, Proteins, Expression, Isolation and Analysis
Order Info
Shipping Condition: Wet Ice
Specifications
Store in refrigerator (2–8°C). | |
10 X | |
3 to 7 | |
Novex™ IEF Gels | |
Wet Ice |
Anode & Cathode Buffers | |
IEF | |
IEF Cathode Buffer | |
Novex™ |
For Research Use Only. Not for use in diagnostic procedures.